Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145401.1 | NP_001138873.1 | 1 | 305 | ||
XM_008758921.1 | XP_008757143.1 | 1 | 650 | |||
XM_008758922.1 | XP_008757144.1 | 1 | 650 | |||
GenBank mRNA | BC161877.1 | - | 1 | 305 |