Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025678.1 | NP_001020849.1 | 2 | 1184 | ||
XM_006228414.2 | XP_006228476.1 | 3 | 3266 | |||
XM_006228415.2 | XP_006228477.1 | 3 | 3269 | |||
GenBank mRNA | BC099170.1 | - | 2 | 1184 | 7382397 |