Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107480.2 | NP_001100950.2 | 12 | 2045 | ||
NM_001161809.1 | NP_001155281.1 | 12 | 2024 | |||
GenBank mRNA | BC168948.1 | - | 11 | 1231 | 7308304 |