Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107482.1 | NP_001100952.1 | 4 | 770 | ||
GenBank mRNA | BC166755.1 | - | 4 | 580 | 7930156 |