Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201562.3 | NP_963856.1 | 9, 10 | 1328 | ||
GenBank mRNA | AY279211.1 | - | 9, 10 | 1328 | ||
BC129079.1 | - | 9, 10 | 1443 | 8370402 | ||
FQ215784.1 | - | 6, 7 | 580 |