Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024258.1 | NP_001019429.1 | 2 | 1129 | ||
XM_008758930.1 | XP_008757152.1 | 2 | 1117 | |||
XM_008758931.1 | XP_008757153.1 | 2 | 1178 | |||
XM_008758932.1 | XP_008757154.1 | 2 | 1163 | |||
GenBank mRNA | BC094953.1 | - | 2 | 1129 | 7308498 |