Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004083.1 | NP_001004083.1 | 3 | 1222 | ||
XM_006228574.2 | XP_006228636.1 | 3 | 1232 | |||
XM_006228575.2 | XP_006228637.1 | 3 | 1271 | |||
GenBank mRNA | AY229997.1 | - | 2 | 952 | ||
AY952204.1 | - | 2 | 952 | |||
BC078907.1 | - | 3 | 1222 | 7113703 | ||
BC081858.1 | - | 3 | 1245 | 7107517 |