Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014018.2 | NP_001014040.2 | 6 | 968 | ||
XM_008759352.1 | XP_008757574.1 | 5 | 2348 | |||
XM_008759353.1 | XP_008757575.1 | 6 | 2414 | |||
XM_008759354.1 | XP_008757576.1 | 6 | 1558 | |||
XM_008759355.1 | XP_008757577.1 | 7 | 1624 | |||
XM_008759356.1 | XP_008757578.1 | 6 | 1981 | |||
XM_008759357.1 | XP_008757579.1 | 6 | 1978 | |||
XM_008759358.1 | XP_008757580.1 | 6 | 1955 | |||
XM_008759359.1 | XP_008757581.1 | 6 | 1946 | |||
XM_008759360.1 | XP_008757582.1 | 6 | 1186 | |||
XM_008759361.1 | XP_008757583.1 | 7 | 1156 | |||
XM_008759362.1 | XP_008757584.1 | 7 | 1084 | |||
XM_008759363.1 | XP_008757585.1 | 7 | 1252 | |||
XM_008759364.1 | XP_008757586.1 | 6 | 1007 | |||
XM_008759365.1 | XP_008757587.1 | 7 | 1075 | |||
GenBank mRNA | BC060592.1 | - | 6 | 972 | 6887297 | |
FQ220903.1 | - | 1 | 972 |