Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175708.3 | NP_783639.1 | 6, 7 | 866 | ||
GenBank mRNA | AY075025.1 | - | 6, 7 | 650 | ||
BC086972.1 | - | 6, 7 | 866 | 7190978 |