Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc90b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024885.1 | NP_001020056.1 | 4 | 713 | ||
GenBank mRNA | BC097480.1 | - | 4 | 713 | 7458611 | |
FQ215669.1 | - | 4 | 415 | |||
FQ216256.1 | - | 4 | 417 | |||
FQ218235.1 | - | 4 | 428 |