Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000504.1 | NP_001000504.1 | 1 | 529 | ||
GenBank mRNA | AF091561.1 | - | 1 | 325 |
Gene Symbol: Olr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000935.1 | NP_001000935.1 | 1 | 529 |