Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082572.1 | NP_001076041.1 | 6 | 1276 | ||
XM_006229884.2 | XP_006229946.1 | 7 | 1424 | |||
XM_006229885.2 | XP_006229947.1 | 7 | 1397 |