Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033998.2 | NP_001029170.1 | 6 | 526 | ||
XM_006230269.2 | XP_006230331.1 | 6 | 667 | |||
GenBank mRNA | AY256462.1 | - | 1 | 29 | ||
BC101849.1 | - | 6 | 526 | 7938065 |