Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003559.4 | NP_003550.1 | 3 | 764 | ||
XM_011525326.2 | XP_011523628.1 | 3 | 824 | |||
XM_011525327.2 | XP_011523629.1 | 4 | 343 | |||
XM_011525329.2 | XP_011523631.1 | 3 | 824 | |||
XM_017025196.1 | XP_016880685.1 | 4 | 341 | |||
XM_017025197.1 | XP_016880686.1 | 2 | 74 | |||
XM_017025199.1 | XP_016880688.1 | 3 | 174 | |||
GenBank mRNA | BC027459.1 | - | 3 | 381 | 3877995 | |
BT019614.1 | - | 3 | 283 | |||
U85245.1 | - | 3 | 763 |