Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exnef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079706.1 | NP_001073174.1 | 7 | 722 | ||
XM_006230111.2 | XP_006230173.1 | 6 | 652 | |||
XM_006230121.2 | XP_006230183.1 | 3 | 604 | |||
XM_008759751.1 | XP_008757973.1 | 6 | 658 | |||
XM_008759752.1 | XP_008757974.1 | 6 | 943 | |||
XM_008759753.1 | XP_008757975.1 | 6 | 944 | |||
XM_008759754.1 | XP_008757976.1 | 6 | 658 | |||
XM_008759755.1 | XP_008757977.1 | 6 | 943 | |||
XM_008759756.1 | XP_008757978.1 | 6 | 943 | |||
XM_008759757.1 | XP_008757979.1 | 4 | 703 | |||
XM_008759758.1 | XP_008757980.1 | 6 | 945 | |||
XM_008759759.1 | XP_008757981.1 | 5 | 431 | |||
XM_008759760.1 | XP_008757982.1 | 6 | 658 | |||
GenBank mRNA | BC128775.1 | - | 7 | 722 | 7133646 |