Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003561.2 | NP_003552.1 | 3, 4 | 706 | ||
GenBank mRNA | BC069539.1 | - | 3, 4 | 546 | 7262248 | |
BC106731.1 | - | 3, 4 | 706 | 40015660 | ||
BC106732.1 | - | 3, 4 | 706 | 40015664 | ||
BC111804.1 | - | 3, 4 | 706 | 40069347 | ||
CR456885.1 | - | 3, 4 | 362 | |||
U95301.1 | - | 3, 4 | 802 |