Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPARCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128310.2 | NP_001121782.1 | 8 | 2011 | ||
NM_001291976.1 | NP_001278905.1 | 8 | 2018 | |||
NM_001291977.1 | NP_001278906.1 | 5 | 1669 | |||
NM_004684.5 | NP_004675.3 | 7 | 1909 | |||
GenBank mRNA | AK294335.1 | - | 8 | 1366 | ||
AK298039.1 | - | 8 | 1182 | |||
AK300956.1 | - | 8 | 1318 | |||
AK304494.1 | - | 5 | 1538 | |||
AK304628.1 | - | 7 | 1151 | |||
AK316533.1 | - | 8 | 1608 | |||
AL833430.1 | - | 7 | 1647 | |||
AY513278.1 | - | 7 | 1519 | |||
BC033721.1 | - | 7 | 1684 | 4939390 | ||
BX647713.1 | - | 8 | 1560 | |||
X82157.1 | - | 7 | 1636 | |||
X86693.1 | - | 7 | 1761 |