Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007226.1 | NP_001007227.1 | 11, 12 | 1454 | ||
NM_001007227.1 | NP_001007228.1 | 10, 11 | 1332 | |||
NM_001007228.1 | NP_001007229.1 | 9, 10 | 1280 | |||
NM_001007229.1 | NP_001007230.1 | 10, 11 | 1331 | |||
NM_001007230.1 | NP_001007231.1 | 11, 12 | 1383 | |||
NM_003563.3 | NP_003554.1 | 10, 11 | 1402 | |||
XM_005257723.4 | XP_005257780.1 | 10, 11 | 1626 | |||
XM_005257724.4 | XP_005257781.1 | 9, 10 | 1556 | |||
XM_017025204.1 | XP_016880693.1 | 8, 9 | 1069 | |||
GenBank mRNA | AJ000644.1 | - | 9, 10 | 1140 | ||
AK222589.1 | - | 9, 10 | 1105 | |||
AK293045.1 | - | 10, 11 | 1613 | |||
AK293587.1 | - | 9, 10 | 1086 | |||
AK300616.1 | - | 9, 10 | 1104 | |||
AK303268.1 | - | 8, 9 | 1026 | |||
AK312691.1 | - | 9, 10 | 1116 | |||
AK316366.1 | - | 11, 12 | 1181 | |||
BC001269.1 | - | 10, 11 | 1286 | 3461565 | ||
BC003385.1 | - | 9, 10 | 1097 | 3445878 |