Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177482.1 | NP_803435.1 | 2 | 114 | ||
GenBank mRNA | V01545.1 | - | 2 | 114 |