Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199103.1 | NP_954534.1 | 3, 4 | 526 | ||
XM_006229580.2 | XP_006229642.1 | 5, 6 | 1774 | |||
XM_006229584.2 | XP_006229646.1 | 5, 6 | 760 | |||
XM_006229585.2 | XP_006229647.1 | 4, 5 | 720 | |||
XM_008773600.1 | XP_008771822.1 | 4, 5 | 4834 | |||
XM_008773601.1 | XP_008771823.1 | 5, 6 | 1777 | |||
XM_008773602.1 | XP_008771824.1 | 5, 6 | 1791 | |||
XM_008773603.1 | XP_008771825.1 | 5, 6 | 1782 | |||
XM_008773604.1 | XP_008771826.1 | 4, 5 | 4831 | |||
XM_008773605.1 | XP_008771827.1 | 4, 5 | 4831 | |||
XM_008773606.1 | XP_008771828.1 | 4, 5 | 4828 | |||
XM_008773607.1 | XP_008771829.1 | 5, 6 | 4874 | |||
XM_008773608.1 | XP_008771830.1 | 5, 6 | 764 | |||
XM_008773609.1 | XP_008771831.1 | 5, 6 | 972 | |||
XM_008773610.1 | XP_008771832.1 | 4, 5 | 724 | |||
GenBank mRNA | AY392762.1 | - | 3, 4 | 526 |