Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107587.1 | NP_001101057.1 | 2 | 2425 | ||
XM_006257625.2 | XP_006257687.1 | 3 | 2707 | |||
XM_006257626.2 | XP_006257688.1 | 4 | 2427 | |||
XM_008773586.1 | XP_008771808.1 | 3 | 2718 |