Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107588.1 | NP_001101058.1 | 18 | 2002 | ||
XM_006231481.2 | XP_006231543.1 | 18 | 1999 | |||
XM_008760418.1 | XP_008758640.1 | 18 | 2000 | |||
XM_008760419.1 | XP_008758641.1 | 18 | 2000 | |||
GenBank mRNA | BC168905.1 | - | 18 | 1977 | 9088878 |