Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013153.1 | NP_001013171.1 | 3 | 945 | ||
XM_008760420.1 | XP_008758642.1 | 3 | 432 | |||
XM_008760421.1 | XP_008758643.1 | 3 | 1053 | |||
GenBank mRNA | BC079260.1 | - | 3 | 945 | 7133433 |