Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001176.3 | NP_001167.2 | 6 | 920 | ||
GenBank mRNA | AB593111.1 | - | 6 | 1080 | ||
BC047699.1 | - | 6 | 922 | 5581679 |