Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003568.2 | NP_003559.2 | 11, 12 | 1269 | ||
XM_011510058.2 | XP_011508360.1 | 11, 12 | 988 | |||
GenBank mRNA | AF230929.1 | - | 11, 12 | 1025 | ||
AJ009985.1 | - | 11, 12 | 1214 | |||
AK314926.1 | - | 10, 11 | 1122 | |||
BC005830.2 | - | 10, 11 | 925 | 2959488 | ||
CR536481.1 | - | 9, 10 | 778 |