Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326578.1 | NP_001313507.1 | 10 | 1052 | ||
NM_001326579.1 | NP_001313508.1 | 10 | 839 | |||
NM_003569.2 | NP_003560.2 | 10 | 975 | |||
NR_137169.1 | 9 | 902 | ||||
GenBank mRNA | AK294911.1 | - | 10 | 724 | ||
AK301464.1 | - | 4 | 338 | |||
BC011975.1 | - | 10 | 839 | 3860059 | ||
U77942.1 | - | 10 | 804 |