Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127484.1 | NP_001120956.1 | 2 | 223 | ||
XM_006231508.2 | XP_006231570.1 | 2 | 2218 | |||
XM_008760435.1 | XP_008758657.1 | 2 | 941 | |||
XM_008760436.1 | XP_008758658.1 | 2 | 2218 | |||
GenBank mRNA | BC166754.1 | - | 2 | 160 | 7929564 | |
FQ217301.1 | - | 2 | 356 |