Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107606.1 | NP_001101076.1 | 14 | 2618 | ||
XM_006231513.2 | XP_006231575.1 | 15 | 3035 | |||
XM_006231514.2 | XP_006231576.1 | 14 | 2955 | |||
XM_008760439.1 | XP_008758661.1 | 14 | 2990 |