Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213563.1 | NP_998728.1 | 24 | 2709 | ||
XM_006255932.1 | XP_006255994.1 | 24 | 2762 | |||
XM_006255933.1 | XP_006255995.1 | 24 | 2307 | |||
XM_006255934.1 | XP_006255996.1 | 24 | 2762 |