Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107615.1 | NP_001101085.1 | 5 | 2020 | ||
XM_006256129.2 | XP_006256191.1 | 6 | 2460 | |||
XM_006256130.2 | XP_006256192.1 | 6 | 2460 | |||
XM_006256131.2 | XP_006256193.1 | 6 | 2169 | |||
XM_006256132.2 | XP_006256194.1 | 6 | 2377 |