Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107631.1 | NP_001101101.1 | 7 | 1222 | ||
GenBank mRNA | BC168160.1 | - | 7 | 1251 | 8364776 |