Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013156.1 | NP_001013174.1 | 2, 3 | 576 | ||
XM_006231841.2 | XP_006231903.1 | 3, 4 | 678 | |||
GenBank mRNA | BC083695.1 | - | 2, 3 | 576 | 7189545 |