Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144772.1 | NP_001138244.1 | 17, 18 | 1478 | ||
NM_003580.3 | NP_003571.2 | 17, 18 | 1538 | |||
GenBank mRNA | AK292679.1 | - | 17, 18 | 1418 | ||
AK294009.1 | - | 17, 18 | 1478 | |||
AK313991.1 | - | 17, 18 | 1449 | |||
BC041124.1 | - | 17, 18 | 1520 | 5785460 | ||
X96586.1 | - | 17, 18 | 1336 |