Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107647.2 | NP_001101117.1 | 5 | 638 | ||
XM_006231836.2 | XP_006231898.1 | 5 | 1066 | |||
XM_008760671.1 | XP_008758893.1 | 4 | 3261 | |||
XM_008760672.1 | XP_008758894.1 | 5 | 3113 | |||
XM_008760673.1 | XP_008758895.1 | 5 | 870 | |||
GenBank mRNA | BC131851.1 | - | 5 | 638 | 7189311 | |
FQ211666.1 | - | 5 | 689 |