Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749226.3 | XP_003749274.1 | 27, 28 | 4738 | ||
XM_006232090.2 | XP_006232152.1 | 27, 28 | 4738 | |||
XM_006232091.2 | XP_006232153.1 | 27, 28 | 4613 |