Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007598.3 | NP_001007599.3 | 5 | 387 | ||
GenBank mRNA | BC060589.1 | - | 5 | 391 | 6888030 | |
BC086561.1 | - | 4 | 369 | 7304371 | ||
FQ210154.1 | - | 5 | 387 | |||
FQ210235.1 | - | 5 | 383 | |||
FQ211371.1 | - | 5 | 387 | |||
FQ212163.1 | - | 5 | 386 | |||
FQ212174.1 | - | 5 | 387 | |||
FQ212384.1 | - | 5 | 391 | |||
FQ212431.1 | - | 5 | 386 | |||
FQ216865.1 | - | 5 | 387 | |||
FQ220825.1 | - | 5 | 385 | |||
FQ221003.1 | - | 5 | 387 | |||
FQ221028.1 | - | 5 | 387 | |||
FQ221603.1 | - | 5 | 387 | |||
FQ221794.1 | - | 5 | 389 | |||
FQ221999.1 | - | 5 | 389 | |||
FQ222130.1 | - | 5 | 389 | |||
FQ222146.1 | - | 5 | 387 | |||
FQ222204.1 | - | 5 | 387 | |||
FQ222278.1 | - | 5 | 385 | |||
FQ222508.1 | - | 5 | 389 | |||
FQ222509.1 | - | 5 | 387 | |||
FQ222609.1 | - | 5 | 389 | |||
FQ222630.1 | - | 5 | 386 | |||
FQ222743.1 | - | 5 | 387 | |||
FQ223008.1 | - | 5 | 401 | |||
FQ223059.1 | - | 5 | 387 | |||
FQ223395.1 | - | 5 | 389 | |||
FQ223420.1 | - | 5 | 385 | |||
FQ223651.1 | - | 5 | 382 | |||
FQ224555.1 | - | 5 | 387 | |||
FQ228320.1 | - | 5 | 388 | |||
FQ228532.1 | - | 5 | 399 | |||
FQ229128.1 | - | 5 | 385 | |||
X51706.1 | - | 4 | 369 |
Gene Symbol: LOC100360449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729147.2 | XP_002729193.1 | 1 | 393 |
Gene Symbol: LOC100364457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748774.3 | XP_003748822.1 | 1 | 400 |
Gene Symbol: LOC103690431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768786.1 | XP_008767008.1 | 2 | 435 |
Gene Symbol: LOC103690796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773059.1 | XP_008771281.1 | 366 |
Gene Symbol: LOC103693978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771314.1 | XP_008769536.1 | 1 | 336 |