Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107678.1 | NP_001101148.1 | 7, 8 | 1031 | ||
XM_008761047.1 | XP_008759269.1 | 7, 8 | 1032 |