Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013158.1 | NP_001013176.1 | 3 | 351 | ||
XM_008761089.1 | XP_008759311.1 | 1 | 195 | |||
GenBank mRNA | BC079206.1 | - | 3 | 351 | 7131789 |