Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 3 | 477 | ||
NM_001278513.1 | NP_001265442.1 | 3 | 621 | |||
NM_001278514.1 | NP_001265443.1 | 3 | 265 | |||
NM_003589.2 | NP_003580.1 | 3 | 303 | |||
XM_011537523.2 | XP_011535825.1 | 2 | 837 | |||
GenBank mRNA | AB178950.1 | - | 3 | 354 | ||
AF077188.1 | - | 3 | 214 | |||
AK314835.1 | - | 3 | 576 | |||
AL833355.1 | - | 2 | 217 | |||
AY365124.1 | - | 3 | 354 | |||
BC008308.1 | - | 3 | 365 | 3537176 | ||
BX439434.2 | - | 3 | 306 | |||
DA574202.1 | - | 3 | 265 | |||
DB455959.1 | - | 3 | 477 |