Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 6 | 667 | ||
NM_001278513.1 | NP_001265442.1 | 6 | 811 | |||
NM_001278514.1 | NP_001265443.1 | 6 | 479 | |||
NM_003589.2 | NP_003580.1 | 6 | 493 | |||
XM_011537523.2 | XP_011535825.1 | 5 | 1027 | |||
GenBank mRNA | AB178950.1 | - | 6 | 544 | ||
AF077188.1 | - | 6 | 404 | |||
AK126278.1 | - | 1 | 876 | |||
AK314835.1 | - | 6 | 766 | |||
AL833355.1 | - | 5 | 431 | |||
AY365124.1 | - | 6 | 544 | |||
BC008308.1 | - | 6 | 555 | 3537176 | ||
DA574202.1 | - | 6 | 455 |