Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOX2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005169.3 | NP_005160.2 | 3 | 1482 | ||
GenBank mRNA | BC041564.1 | - | 3 | 1464 | 5923445 |