Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107691.1 | NP_001101161.1 | 3 | 405 | ||
XM_008761170.1 | XP_008759392.1 | 3 | 522 | |||
XM_008761171.1 | XP_008759393.1 | 3 | 369 | |||
XM_008761172.1 | XP_008759394.1 | 3 | 368 | |||
XM_008761173.1 | XP_008759395.1 | 3 | 369 |