Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207605.3 | NP_997488.2 | 7 | 1001 | ||
GenBank mRNA | AF468787.2 | - | 7 | 993 | ||
BC088087.1 | - | 7 | 1001 | 7379832 |