Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107710.1 | NP_001101180.1 | 14 | 2159 | ||
XM_006233083.2 | XP_006233145.1 | 14 | 2554 |