Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107714.1 | NP_001101184.1 | 11 | 1339 | ||
XM_006233114.2 | XP_006233176.1 | 11 | 1288 | |||
XM_008761396.1 | XP_008759618.1 | 10 | 1735 | |||
XM_008761397.1 | XP_008759619.1 | 10 | 1735 | |||
XM_008761398.1 | XP_008759620.1 | 11 | 1288 | |||
XM_008761399.1 | XP_008759621.1 | 11 | 1372 | |||
GenBank mRNA | BC158880.1 | - | 11 | 1275 | 7936253 |