Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014042.1 | NP_001014064.1 | 2, 3 | 364 | ||
XM_006233150.2 | XP_006233212.1 | 2, 3 | 730 | |||
GenBank mRNA | BC087113.1 | - | 2, 3 | 364 | 7193587 |