Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233151.2 | XP_006233213.1 | 4 | 371 | ||
GenBank mRNA | BC166866.1 | - | 4 | 335 | 8369659 | |
CB557548.1 | - | 3 | 197 | |||
FM135814.1 | - | 4 | 309 |