Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107720.1 | NP_001101190.1 | 3 | 1033 | ||
XM_008761462.1 | XP_008759684.1 | 3 | 2240 | |||
GenBank mRNA | EU792473.1 | - | 3 | 1033 |