Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107725.1 | NP_001101195.1 | 8 | 1632 | ||
XM_006233324.2 | XP_006233386.1 | 8 | 1713 | |||
XM_008761502.1 | XP_008759724.1 | 7 | 2466 | |||
XM_008761503.1 | XP_008759725.1 | 8 | 3289 | |||
XM_008761504.1 | XP_008759726.1 | 8 | 3284 | |||
XM_008761505.1 | XP_008759727.1 | 8 | 3247 | |||
GenBank mRNA | FQ211741.1 | - | 8 | 1718 |
Gene Symbol: LOC102556234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233344.2 | XP_006233406.1 | 308 |