Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107725.1 | NP_001101195.1 | 2 | 223 | ||
XM_006233324.2 | XP_006233386.1 | 2 | 304 | |||
XM_008761502.1 | XP_008759724.1 | 1 | 1057 | |||
XM_008761503.1 | XP_008759725.1 | 2 | 1880 | |||
XM_008761504.1 | XP_008759726.1 | 2 | 1875 | |||
XM_008761505.1 | XP_008759727.1 | 2 | 1838 | |||
GenBank mRNA | FQ211741.1 | - | 2 | 309 |